Modifying Enzymes
Thermo Scientific™

Modifying Enzymes

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantityConcentration
4387661AFT4 RNA Ligase 21000 U10 U/μL
EM167AFVaccinia mRNA Cap 2'-O-Methyltransferase20,000 U50 U/μL
EO166AFVaccinia Capping Enzyme4000 U10 U/μL
EL002AFT4 RNA Ligase1500 U30 U/μL
EN230AFDNase I5000 U50 U/μL
Catalog number 4387661AF
Price (INR)
22,947.00
線上優惠
Ends: 30-Sep-2026
41,721.00
Save 18,774.00 (45%)
Each
Product Type:
T4 RNA Ligase 2
Quantity:
1000 U
Concentration:
10 U/μL
Price (INR)
22,947.00
線上優惠
Ends: 30-Sep-2026
41,721.00
Save 18,774.00 (45%)
Each

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer10T T4 RNA Ligase 2 Reaction Buffer
For Use With (Application)RNA Ligation
InhibitorsEDTA
Product TypeT4 RNA Ligase 2
Quantity1000 U
Concentration10 U/μL
FormLiquid
Research CategoryRNA Research
Sensitive to Heat InactivationNo
Unit SizeEach
Contents & Storage
10 U/μL T4 RNA Ligase 2, 100 μL

Store at -20°C.