O-Phenylhydroxylamine hydrochloride, 97%
O-Phenylhydroxylamine hydrochloride, 97%
O-Phenylhydroxylamine hydrochloride, 97%
Acros Organics

O-Phenylhydroxylamine hydrochloride, 97%

CAS: 6092-80-4 | C6H7NO·ClH | 145.59 g/mol
Have Questions?
Catalog NumberQuantityPackaging
438810010
also known as 43881
1 gGlass bottle
Catalog number 438810010
also known as 43881
HSN 29280090
Price (INR)
27,932.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
27,932.00
Each
Chemical Identifiers
CAS6092-80-4
IUPAC Namehydrogen O-phenylhydroxylamine chloride
Molecular FormulaC6H8ClNO
InChI KeyDBTXKJJSFWZJNS-UHFFFAOYSA-N
SMILES[H+].[Cl-].NOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink to brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only