Product Image
Applied Biosystems™

SOLiD™ Deposition Chambers, 1 well

Have Questions?
Catalog NumberQuantity
43928382 each
Catalog number 4392838
Price (INR)
-
Quantity:
2 each
This is an accessory for use with the SOLiD™ Analyzer.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)SOLiD™ 3 Analyzer, SOLiD™ 4 System
Format1-well
Product TypeDeposition Chamber
Quantity2 each
Shipping ConditionRoom Temperature
Product LineSOLiD
Unit Size2 each
Contents & Storage
Store at room temperature. Includes two additional deposition chambers for 1 sample⁄slide.