Product Image
Applied Biosystems™

PrepFiler™ 96-Well Processing Plates

Have Questions?
Catalog NumberQuantity
439290410 plates
Catalog number 4392904
Price (INR)
27,435.00
Each
Quantity:
10 plates
Price (INR)
27,435.00
Each
The PrepFiler 96-well Processing Plate is now available for purchase separately to provide the ordering flexibility to select only the plastics consumables needed for use with the PrepFiler Automated Forensic DNA Extraction Kit. The full PrepFiler Automated Forensic DNA Extraction Kit uses magnetic particles with an optimized multi-component surface chemistry to deliver superior DNA yield from most forensic sample types, including body fluids, and stains and swabs of body fluids.

The PrepFiler 96-well Processing Plate is used for sample processing in 96-well format.

The full PrepFiler Automated Forensic DNA Extraction Kit is also available for purchase, and contains other components required for DNA recovery from both routine and challenging samples, including removal of PCR inhibitors.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)PrepFiler
Plate TypeReceiver Plate
Product LinePrepFiler
Product Type96-Well Processing Plate
Quantity10 plates
Shipping ConditionRoom Temperature
Format96-well Plate
Unit SizeEach
Contents & Storage
Store plates at ambient temperature.