6-Quinolineboronic acid pinacol ester, 97%
6-Quinolineboronic acid pinacol ester, 97%
6-Quinolineboronic acid pinacol ester, 97%
Acros Organics

6-Quinolineboronic acid pinacol ester, 97%

CAS: 406463-06-7 | C15H18BNO2 | 255.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 439640050
also known as 43964
HSN 29334900
Price (INR)
47,651.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
47,651.00
Each
Chemical Identifiers
CAS406463-06-7
IUPAC Name6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular FormulaC15H18BNO2
InChI KeyVMFALDWCEQUFSX-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC2=CC=CN=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point62°C to 70°C
GC>=96.0 %
Appearance (Color)White to light orange
Appearance (Form)Powder or crystalline solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only