Product Image
Applied Biosystems™
GeneMapper™ ID Software v3.3 Initial License
GeneMapper™ ID Software version 3.3 (Simplified Chinese Version) and the AmpFLSTR™ Sinofiler™ PCR Amplification Kit are designed specifically to meettheRead more
Have Questions?
Catalog NumberQuantity
43981761 license
Catalog number 4398176
Price (INR)
2,280,681.00
Each
Quantity:
1 license
Price (INR)
2,280,681.00
Each
GeneMapper™ ID Software version 3.3 (Simplified Chinese Version) and the AmpFLSTR™ Sinofiler™ PCR Amplification Kit are designed specifically to meetthe needs of Chinese forensic DNA laboratories. When used together as part of an integrated solution, GeneMapper™ ID Software version 3.3 and the AmpFLSTR™ Sinofiler™ PCR Amplification Kit provide the necessary functionality and flexibility to address the diverse challenges faced when processing forensic casework and high volume convicted offender DNA samples.
• Contains new version (v2_33) of panel and binset files that support the new AmpFLSTR™ Sinofiler™ PCR Amplification Kit, as well as previously supported AmpFLSTR™ kits
• Automatically imports the panel and binset files and eliminates the need to manually import the panel and binset
• All static user interfaces translated in Simplified Chinese
• GeneMapper™ ID Software v3.3 has the same analysis algorithms as GeneMapper™ ID Software v3.2.1
Reduce Analysis Bottlenecks
The software's novel Process Component-Based Quality Values (PQVs) enable you to identify and sort genotypes quickly based on user-defined quality thresholds. Check the concordance of overlapping controls and loci from different AmpFLSTR™ kits automatically -- significantly improving analysis workflow and productivity.
A Complete, Verified System
GeneMapper™ ID Software v3.2.1 is an integral component of Applied Biosystems complete system of instruments, reagents, software, and service for human-identity applications. It has undergone extensive verification testing to ensure robust design features, performance, and reproducibility you can depend on.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntryGenetic Analyzer Data
Data OutputsProcess-Component-Based Quality Value (PQV) reports
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer, 3700 Genetic Analyzer, 3700 Sequencer, 377 DNA Sequencer
LicenseStatic
Operating SystemWindows 2000 Pro (Service Pack 3), Windows XP Pro (Service Pack 2)
Product TypeSoftware V3.3
Quantity1 license
Software CategorySoftware
Product LineGeneMapper
Software TypeGenotyping, GeneMapper
Starting MaterialDNA
Unit SizeEach