The AmpliTaq Gold 360 Buffer Kit is used in conjunction with AmpliTaq Gold 360 DNA Polymerase and is intended for those occasions when the quantity of reagents supplied with the enzyme are not sufficient.
Have Questions?
Catalog Number
Quantity
4398853
1 Kit
Catalog number 4398853
Price (INR)
-
Quantity:
1 Kit
Request bulk or custom format
The AmpliTaq Gold 360 Buffer Kit is used in conjunction with AmpliTaq Gold 360 DNA Polymerase and is intended for use on occasions when the quantity of reagents supplied with the enzyme are not sufficient.
This kit includes 1.5 mL (each) of:
10X AmpliTaq Gold 360 Buffer
25 mM MgCl2
360 GC Enhancer
Typically, 1.5 mL is sufficient for use with 250 U of enzyme.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)1X
FormatKit
PolymeraseAmpliTaq Gold 360 DNA Polymerase
Product TypePCR Buffer
Quantity1 Kit
Shipping ConditionRoom Temp or Wet Ice
Volume1.5 mL
Concentration10X
For Use With (Application)Cloning
Unit SizeEach
Contents & Storage
• 10X AmpliTaq Gold 360 Buffer, 1 x 1.5 mL • 25 mM MgCl2, 1 x 1.5 mL • 360 GC Enhancer, 1 x 1.5 mL
Store at -15° to -25°C.
Frequently asked questions (FAQs)
My oligonucleotide does not appear to be the right length when I checked by gel electrophoresis. Why is this?
Oligos should be run on a polyacrylamide gel containing 7 M urea and loaded with a 50% formamide solution to avoid compressions and secondary structures. Oligos of the same length and different compositions can electrophorese differently. dC's migrate fastest, followed by dA's, dT's, and then dG's. Oligos containing N's tend to run as a blurry band and generally have a problem with secondary structure.
The primers I am using worked for PCR initially, but over time, have stopped working. What happened?
Primers should be aliquoted for single use before PCR set-up. Heat just the aliquoted primers to 94 degrees for 1 min. Quick chill the primer on ice before adding to the PCR reaction. Some primers may anneal to themselves or curl up on themselves.
I don't see a pellet in my oligo tube order. Should I ask for a replacement?
The drying method dries the primer in a thin layer along the sidewalls of the tube instead of the bottom, therefore a pellet is not always visible and should still be ready to use.
There is a ball-shaped pellet at the bottom of my oligo tube. What is this and can I still use my oligo?
If the oligo was overheated, it will appear as a ball-shaped pellet attached to the bottom of the tube. This should not affect the quality of the oligo, and the oligo should be readily soluble in water.
There is a green color in my lyophilized oligo. Can I still use it?
If an oligo appears green in color, this is most likely due to ink falling into the tube. The oligo should still be fully functional. The color can be removed by doing an ethanol precipitation.