Cell Fractionation Buffer for PrepSEQ™ Nucleic Acid Sample Preparation Kit
Applied Biosystems™

Cell Fractionation Buffer for PrepSEQ™ Nucleic Acid Sample Preparation Kit

Cell Fractionation Buffer is used to process the mammalian cell pellet to obtain free Mycoplasma and then combine with the Mycoplasma pellet in the large scale PrepSEQ extraction protocol.
Have Questions?
Catalog NumberQuantity
44034613 bottles, 25 mL
Catalog number 4403461
Price (INR)
-
Quantity:
3 bottles, 25 mL

Cell Fractionation Buffer is used to process the mammalian cell pellet to obtain free Mycoplasma and then combine with the Mycoplasma pellet in the large scale PrepSEQ extraction protocol. This protocol can process up to 15 mL of sample (up to 106 cells/mL) for the detection of Mycoplasma.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialCell Fractionation Buffer
Recommended StorageSufficient for 15 mL of sample (up to 106 cells/mL)

Store at 2°C to 8°C.
For Use With (Application)Mycoplasma detection
For Use With (Equipment)PrepSEQ™ Nucleic Acid Sample Preparation Kit
Physical FormLiquid
Quantity3 bottles, 25 mL
Unit SizeEach