3500 Dx Genetic Analyzer Capillary Array, 50 cm
Applied Biosystems™
3500 Dx Genetic Analyzer Capillary Array, 50 cm
A radio-frequency identification tag is built into the frame for tracking critical run information, which is then reported on the software dashboard.
Have Questions?
Catalog NumberQuantity
44046841 Array
Catalog number 4404684
Price (INR)
347,428.00
Each
Quantity:
1 Array
Price (INR)
347,428.00
Each
These internally uncoated 50-cm capillaries are supplied in assemblies of 8 capillaries per array, with a built-in frame for easy installation. A radio-frequency identification (RFID) tag is built into the frame for tracking critical run information, which is then reported on the software dashboard. The array can be used for 160 injections in Dx mode.
This product is intended for in vitro diagnostic use on the 3500 Dx Genetic Analyzer and is not validated for use with other systems or applications. Refer to the 3500 Dx Genetic Analyzer User Guide for further details.
These internally uncoated 50cm capillaries are supplied in preassembled set assemblies of 8 capillaries per array, with a built-in frame for easy installation.
For In Vitro Diagnostic Use Only
Specifications
Capillary Length50 cm
For Use With (Application)Sequencing
For Use With (Equipment)3500 Dx Genetic Analyzer, 3500 Genetic Analyzer
Product Type3500 Dx Genetic Analyzer
Quantity1 Array
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Ambient, 1 array with 8 capillaries