Product Image
Applied Biosystems™
MicroSEQ™ ID Sequencing Clean-up Cartridges
The MicroSEQ ID Sequencing Clean-up Cartridges are used for post-sequencing clean-up in the MicroSEQ ID workflow.
Have Questions?
Catalog NumberQuantity
440822836 cartridges
Catalog number 4408228
Price (INR)
16,172.00
Each
Quantity:
36 cartridges
Price (INR)
16,172.00
Each
The MicroSEQ™ ID Sequencing Clean-up Cartridges are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrix used in the cartridges provides a convenient and simple way to remove dye terminators, dNTPs, and salts, as well as residual primers from cycle sequencing reactions.
The MicroSEQ™ ID Sequencing Clean-up Cartridges can be used downstream of the following sequencing kits:
  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit
The kit consists of 36 cartridges and is ideal for low-throughput microbial identification workflows. The end products can be directly used for sequencing on any of Applied Biosystems™ genetic analyzers.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeDNA
For Use With (Application)Sequencing
For Use With (Equipment)MicroSEQ™ ID
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product LineMicroSEQ
PuritySequencing Grade
Quantity36 cartridges
Shipping ConditionWet Ice
TargetSequencing Reactions
FormatCartridge, Spin Column
Isolation TechnologySpin Column
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).