OpenArray™ Case Sealing Station
Applied Biosystems™

OpenArray™ Case Sealing Station

This is a Case Sealing Station for the OpenArray™ Real-Time PCR Platform.
Have Questions?
Catalog NumberQuantity
44093611 unit
Catalog number 4409361
Price (INR)
-
Quantity:
1 unit

This is a Case Sealing Station for the OpenArray™ Real-Time PCR Platform.

The OpenArray™ Real-Time PCR Instrument and OpenArray™ AccuFill™ System combination can process hundreds to thousands of samples per day, enabling mid-density, high-throughput results with ease of use.

OpenArray™ Real-Time PCR Components
The platform includes the OpenArray™ Real-Time PCR Instrument, computer⁄monitor, analysis software for genotyping and real-time applications, installation, training and OpenArray™ Real-Time PCR Platform warranty.

The OpenArray™ Real-Time PCR Platform includes:

  • OpenArray™ AccuFill™ System—Accurately loads samples onto OpenArray™ Plates
  • OpenArray™ Case Sealing Station—Seals OpenArray™ Plates in cases
  • OpenArray™ Real-Time PCR Software and OpenArray™ Genotyping Software—Acquire, analyze, and manage data quickly and easily

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)OpenArray™
FormatOpenArray™ Plate
Product LineOpenArray
Quantity1 unit
Product TypeCase Sealing Station
Unit SizeEach
Contents & Storage
Store at Room Temperature