Product Image
Applied Biosystems™

Conditioning Reagent Dx

Conditioning Reagent Dx is designed for use with Applied Biosystems Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
44095431 Pk.
Catalog number 4409543
Price (INR)
8,391.00
Each
Quantity:
1 Pk.
Price (INR)
8,391.00
Each

This Conditioning Reagent Dx is designed for use withApplied Biosystems Dx genetic analyzers.

Conditioning Reagent Dx is available as a ready-to-use pouch and is used for priming the polymer pump, washing the polymer pump between polymer type changes, and during instrument shut down. The pouch has adequate volume for a one-time use. The top part of the pouch fitment is sealed with a plastic film, which should be removed prior to installation on to the instrument.

This product is intended for in vitro diagnostic use on Applied Biosystems Dx genetic analyzers and is not validated for use with other systems or applications. Applied Biosystems Dx genetic analyzers are to be used by trained operators in the analysis of human DNA for the detection of fluorescently labeled DNA nucleotides by capillary electrophoresis. Refer to the appropriate Applied Biosystems Dx genetic analyzer user guide for further details.

The 3500 Dx series conditioning reagent, anode buffer, cathode buffer and septa cathode buffer are required for instrument operation.
For In Vitro Diagnostic Use Only
Specifications
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
FormatPouch
Product TypeConditioning Reagent
Quantity1 Pk.
Shipping ConditionWet Ice
Starting MaterialDNA, RNA
Unit SizeEach
Contents & Storage
Store at 2 - 8°C.