3500 Dx Series 96-Well Fast (0.1 mL) Retainer & Base Set
Applied Biosystems™

3500 Dx Series 96-Well Fast (0.1 mL) Retainer & Base Set

This is a 96-well retainer & base set (fast) designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
44102294 Sets
Catalog number 4410229
Price (INR)
25,100.00
Each
Quantity:
4 Sets
Price (INR)
25,100.00
Each

This is a retainer and base set (fast) designed for septa retention in 8-tube strip and 96-well plate formats for use with the 3500 Dx and 3500xL Dx genetic analyzers.

Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only.

  • This is a 96-well retainer & base set (fast) designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
For In Vitro Diagnostic Use Only
Specifications
Compatibility96-Well Microtiter Plates, MicroAmp™ Optical 96-Well Reaction Plates
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Format96-well
Product TypeRetainer and Base Set
Quantity4 Sets
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Ambient storage.
Each package contains 4 sets.