3500 Dx Series 384-Well Retainer & Base Set
Applied Biosystems™

3500 Dx Series 384-Well Retainer & Base Set

Designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
44102344 Sets
Catalog number 4410234
Price (INR)
26,529.00
Each
Quantity:
4 Sets
Price (INR)
26,529.00
Each

This is a retainer and base set (standard) designed for septa retention in 384-well format for use with the 3500 Dx and 3500xL Dx genetic analyzers.

Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only. Not for use in the USA.

  • This is a 384-well retainer & base set (standard) designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer.
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
For In Vitro Diagnostic Use Only
Specifications
Compatibility384-Well Microtiter Plates, MicroAmp™ Optical 384-Well Reaction Plates
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Format384-well
Product TypeRetainer and Base Set
Quantity4 Sets
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Ambient storage.
Each package contains 4 sets.