3500 Dx Series 8-Strip Septa
Applied Biosystems™

3500 Dx Series 8-Strip Septa

This is an 8-strip septa designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
441070224 Pk.
Catalog number 4410702
Price (INR)
55,253.00
Each
Quantity:
24 Pk.
Price (INR)
55,253.00
Each

This is an 8-strip septa designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.

Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only. Not for use in the USA.

  • This is an 8-strip septa designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only. Not for use in the USA.
For In Vitro Diagnostic Use Only
Specifications
For Use With (Equipment)3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Format8-tube Strip
Product TypeSepta Strip
Quantity24 Pk.
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Ambient storage.
Each package contains 24 sets.