The USDA licensed, VetMAX™ -Gold SIV Detection Kit is a robust PCR diagnostic kit, enabling early detection of swine influenzaRead more
Have Questions?
Catalog Number
Quantity
4415200
100 reactions
Catalog number 4415200
Price (INR)
77,732.00
Each
Quantity:
100 reactions
Price (INR)
77,732.00
Each
The USDA licensed, VetMAX™ -Gold SIV Detection Kit is a robust PCR diagnostic kit, enabling early detection of swine influenza virus in pigs and helping to minimize the spread of the disease.
Licensed by the USDA
• Targets multiple genomic regions, reducing the risk of false negatives caused by genomic mutation • Enables unsurpassed sensitivity, specificity, repeatability, and reproducibility • Decreases variability in results and allows for reproducibility through assay standardization
The VetMAX™-Gold SIV Detection Kit is the first and only USDA-licensed real-time PCR test for the rapid detection of SIV RNA purified from porcine nasal swab samples. This highly sensitive, qualitative, one-step assay delivers accurate results within 1.5 hours. The VetMAX™-Gold SIV Detection assay is a single-tube (or well) real-time RT-PCR assay in which SIV and Xeno™ internal positive control RNA is reverse transcribed into cDNA, then amplified and detected in real time using fluorescent TaqMan™ probes.
Confidence in Results The VetMAX™-Gold SIV Detection Kit utilizes AmpliTaq Gold™ DNA Polymerase and optimized primers and TaqMan™ probes for maximum sensitivity and specificity. Additionally, the included Xeno™ RNA internal positive control enables monitoring of RNA purification efficiency and PCR inhibition, allowing for increased accuracy.
Designed to fit within a simple streamlined workflow, the VetMAX™-Gold SIV Detection Kit is optimized to perform with the MagMAX™ Pathogen RNA/DNA Kit and the Applied Biosystems™ 7500 Fast Real-Time PCR Systems.
Note: This kit is not intended for the differentiation of SIV subtypes.
For Veterinary Use Only
Specifications
Detection MethodPrimer-Probe Detection
For Use With (Application)Sequencing, Real-Time PCR
No. of Reactions100
PCR Method1-Step qRT-PCR
Product LineVetMAX
Product TypeGold SIV Detection Kit
Quantity100 reactions
Sample TypeSwab
Shipping ConditionDry Ice
SpeciesSwine/Porcine
Target Organism ClassSwine Influenza Virus (SIV)
FormatKit
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C; Includes 100 reactions with the following components:
• Multiplex RT-PCR buffer • Multiplex enzyme mix • Nuclease-free water • SIV primer-probe mix • SIV-Xeno RNA control mix • XenoRNA control
Frequently asked questions (FAQs)
What are the quenchers in the VetMAX-Gold SIV Detection Kit?
The VetMAX-Gold SIV Detection Kit contains the following quenchers:
- SIV RNA: Eclipse® Q
- Xeno RNA Control: Eclipse® Q
What are the assay-labeled reporters in the VetMAX-Gold SIV Detection Kit?
The VetMAX-Gold SIV Detection Kit contains the following assay-labeled reporters:
- SIV RNA: FAM dye
- Xeno RNA Control: VIC dye
What is the internal positive control (IPC) in the VetMAX-Gold SIV Detection Kit?
The VetMAX-Gold SIV Detection Kit contains the Xeno RNA Control as the internal positive control (IPC) for the RNA purification process and to monitor for the presence of PCR inhibitors.