VetMAX™ IBR/BHV-1 Reagents
Applied Biosystems™

VetMAX™ IBR/BHV-1 Reagents

Have Questions?
Catalog NumberQuantity
4415203100 reactions
Catalog number 4415203
Price (INR)
62,434.00
Each
Quantity:
100 reactions
Price (INR)
62,434.00
Each
VetMAX™ IBR/BHV-1 Reagents is a TaqMan™ primer probe mix for real-time PCR amplification of Bovine Herpes virus-1 DNA and Xeno™ DNA Internal Positive Control.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions100 Reactions
Product LineVetMAX
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesCattle/Bovine
Sufficient For100 Reactions
TargetBovine Herpesvirus Type 1 (BHV-1)
Target Organism ClassBovine Herpesvirus 1 (BHV-1), Bovine Herpesvirus 1 (BHV-1)
FormLiquid
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Store at -20°C. Includes IBR⁄BHV-1 TaqMan™ Primer Probe Mix.