1-BOC-3-Azetidinone, 97%
1-BOC-3-Azetidinone, 97%
1-BOC-3-Azetidinone, 97%
Acros Organics

1-BOC-3-Azetidinone, 97%

CAS: 398489-26-4 | C8H13NO3 | 171.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 442040050
also known as 44204
HSN 29225090
Price (INR)
42,845.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
42,845.00
Each
Chemical Identifiers
CAS398489-26-4
IUPAC Nametert-butyl 3-oxoazetidine-1-carboxylate
Molecular FormulaC8H13NO3
InChI KeyVMKIXWAFFVLJCK-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Melting point48°C to 54°C
Appearance (Form)Crystalline powder or solid
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only