o-Xylene, 99%, Extra Dry, AcroSeal™
o-Xylene, 99%, Extra Dry, AcroSeal™
o-Xylene, 99%, Extra Dry, AcroSeal™
o-Xylene, 99%, Extra Dry, AcroSeal™
o-Xylene, 99%, Extra Dry, AcroSeal™
Acros Organics

o-Xylene, 99%, Extra Dry, AcroSeal™

CAS: 95-47-6 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 443021000
HSN 29024100
Price (INR)
8,687.00
Each
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (INR)
8,687.00
Each
Add to cart
Chemical Identifiers
CAS95-47-6
IUPAC Name1,2-xylene
Molecular FormulaC8H10
InChI KeyCTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.005 %
Appearance (Color)Clear colorless
Residue after evaporation=<0.01 %
GC>=98.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only