Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
Acros Organics

Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 111-96-6 | C6H14O3 | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Catalog number 443100010
also known as 44310
HSN 29091900
Price (INR)
79,039.00
Each
Quantity:
1 L
Request bulk or custom format
Price (INR)
79,039.00
Each
Chemical Identifiers
CAS111-96-6
IUPAC Name1-methoxy-2-(2-methoxyethoxy)ethane
Molecular FormulaC6H14O3
InChI KeySBZXBUIDTXKZTM-UHFFFAOYSA-N
SMILESCOCCOCCOC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only