VIC™ Dye Calibration Plate, 384-well
VIC™ Dye Calibration Plate, 384-well
Applied Biosystems™

VIC™ Dye Calibration Plate, 384-well

The 384-Well Dye Calibration Plate (VIC®) is used to maintain your real-time PCR instrument with 384-well block. Dye calibration shouldRead more
Have Questions?
Catalog NumberQuantity
44324521 x 384 well plate
Catalog number 4432452
Price (INR)
14,447.00
Each
Add to cart
Quantity:
1 x 384 well plate
Price (INR)
14,447.00
Each
Add to cart
The 384-Well Dye Calibration Plate (VIC®) is used to maintain your real-time PCR instrument with 384-well block. Dye calibration should be run after lamp change. Please see your instrument user guide for full instructions on use.
Specifications
Calibrated DyeVIC
Calibration TypeSpectral Calibration
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)ViiA™ 7 Dx System, QuantStudio™ Dx
Format384-well Dye Calibration Plate
PCR MethodqPCR
Quantity1 x 384 well plate
Shipping ConditionDry Ice
Product TypeDye Calibration Plate
Unit SizeEach
Contents & Storage
Store at –20°C ± 5°C.