ROI & Background Calibration Kit, 384-well
Product Image
Applied Biosystems™

ROI & Background Calibration Kit, 384-well

The 384-Well ROI & Background Calibration Kit is used to maintain your real-time PCR instrument with 384-well block. Region ofRead more
Have Questions?
Catalog NumberQuantity
44324822 x 384 well plates
Catalog number 4432482
Price (INR)
28,179.00
Each
Add to cart
Quantity:
2 x 384 well plates
Price (INR)
28,179.00
Each
Add to cart
The 384-Well ROI & Background Calibration Kit is used to maintain your real-time PCR instrument with 384-well block. Region of interest (ROI) and background calibration should be run after lamp change. Please see your instrument user guide for full instructions on use.
Specifications
Calibrated DyeProprietary Mix
Calibration TypeSpectral Calibration
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)ViiA 7 Dx, QuantStudio Dx, QuantStudio 5 Dx, QuantStudio 7 Pro Dx
PCR MethodqPCR
Quantity2 x 384 well plates
Shipping ConditionDry Ice
Product TypeBackground Calibration Kit
Unit SizeEach
Contents & Storage
Store at –20°C ± 5°C.