Product Image
Applied Biosystems™
Calibration Install Kit, Fast 96-well
This kit is used to maintain your ViiA 7, QuantStudio 6 Flex, QuantStudio 7 Flex, and QuantStudio 12k Flex real-timeRead more
Have Questions?
Catalog NumberQuantity
44325571 kit
Catalog number 4432557
Price (INR)
250,211.00
Each
Quantity:
1 kit
Price (INR)
250,211.00
Each
This kit is used to maintain your ViiA 7, QuantStudio 6 Flex, QuantStudio 7 Flex, and QuantStudio 12k Flex real-time PCR systems with Fast 96-well block. It includes calibration plates to perform a spectral calibration with FAM, VIC, ROX, SYBR Green, TAMRA, NED dyes, plates to perform region-of-interest (ROI) calibration, normalization calibration, and to run an RNase P verification. Calibration and verification should be run at least every six months. Please see your instrument maintenance guide for full instructions on use.
The dye formulations for your real-time PCR system will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibration TypeSpectral Calibration
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)QuantStudio™ 12 K Flex, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
Quantity1 kit
Product TypeCalibration Install Kit
Unit SizeEach
Contents & Storage
Includes:
• 1 background calibration plate
• 1 ROI calibration plate
• 6 spectral calibration plates with FAM, SYBR Green, VIC, NED, TAMRA, and ROX dye standards
• 2 normalization calibration plates
• 1 RNase P instrument verification plate
Store kit at -5 to -30°C.
Frequently asked questions (FAQs)
I cannot open the QuantStudio 3/5 Design & Analysis 2 (DA2) software on my Mac. What are the software requirements for Mac?
To open the software download on your Mac, you will need to adjust your security settings using the following instructions from Apple support:
Safely open apps on your mac
Please note, the software should run on Mac OS 10.10 or later.
Find additional tips, troubleshooting help, and resources within our QuantStudio 3 and QuantStudio 5 Real Time PCR Systems Support Center.
How can I get a summary of the calibrations performed on the QuantStudio 6 and 7 Flex Real-Time PCR Systems?
You can get a summary of the calibrations from the QuantStudio Software by going to Instrument > Instrument Console > Manage Instrument > Information > Print Calibration Log.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Do I need to back up my calibration files before I upgrade the software for the ViiA7 Real-Time PCR System?
No, you do not need to back up the calibration files as all of the calibration files are stored on the ViiA7 Real-Time PCR instrument. Once the computer software is properly installed and the computer is connected to the instrument, the calibration information will automatically update in the QuantStudio Software.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I still start a run on the ViiA7 Real-Time PCR System if the "Calibration is expired" notification is displayed?
Yes, you can still start a run on the ViiA7 Real-Time PCR System if the expired calibration notice displays. This is just a reminder and you can close it to proceed with the run. Additionally, you can change when you want the notifications to show up and how frequently you want to be notified to calibrate the instrument. To do this, go to the toolbar in the QuantStudio Software and select Instrument > Instrument Console. Select your instrument icon and click on Manager Instrument > Maintenance > Calibration Reminders. From the Calibration reminders Settings page you can update when the calibrations expire, turn expiration reminders on/off, and set when the reminders show up.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.