The 96-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with Standard 96-Well Block. Confirm the performanceRead more
Have Questions?
Catalog Number
Quantity
4432593
1 x 96-well plate
Catalog number 4432593
Price (INR)
93,840.00
Each
Add to cart
Quantity:
1 x 96-well plate
Price (INR)
93,840.00
Each
Add to cart
The 96-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with Standard 96-Well Block. Confirm the performance of your instrument by running the RNaseP assay. Please see your instrument user guide for full instructions on use.
The 96-Well RNaseP Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.
The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.
Specifications
Calibration TypeInstrument Verification Plate
For Use With (Equipment)ViiA 7 Dx, QuantStudio Dx, QuantStudio 5 Dx, QuantStudio 7 Pro Dx