Normalization Kit, 384-well
Product Image
Applied Biosystems™

Normalization Kit, 384-well

The 384-Well Normalization Kit is used to maintain your real-time PCR instrument with 384-well block. Normalization plates are used toRead more
Have Questions?
Catalog NumberQuantity
44326202 x 384-well plates
Catalog number 4432620
Price (INR)
25,336.00
Each
Add to cart
Quantity:
2 x 384-well plates
Price (INR)
25,336.00
Each
Add to cart
The 384-Well Normalization Kit is used to maintain your real-time PCR instrument with 384-well block. Normalization plates are used to calibrate different instruments. The kit contains one FAM™/ROX™ normalization plate and one VIC®/ROX™ normalization plate. Please see your instrument user guide for full instructions on use.
Specifications
Calibrated DyeProprietary Mix
Calibration TypeSpectral Calibration
For Use With (Application)Real Time PCR (qPCR)
For Use With (Equipment)ViiA™ 7 Dx System, QuantStudio™ Dx
PCR MethodqPCR
Quantity2 x 384-well plates
Shipping ConditionDry Ice
Product TypeNormalization Kit
Unit SizeEach
Contents & Storage
Store at –20°C ± 5°C.