2-Amino-5-methoxybenzoic acid, 97%
2-Amino-5-methoxybenzoic acid, 97%
2-Amino-5-methoxybenzoic acid, 97%
Acros Organics

2-Amino-5-methoxybenzoic acid, 97%

CAS: 6705-03-9 | C11H13N3O | 203.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 443340010
also known as 44334
HSN 29225090
Price (INR)
9,279.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,279.00
Each
Chemical Identifiers
CAS6705-03-9
IUPAC Name2-amino-5-methoxybenzoic acid
Molecular FormulaC8H9NO3
InChI KeyUMKSAURFQFUULT-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or solid
HPLC>=96.0 %
Appearance (Color)Off-white to brown to purple
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only