3500xL Genetic Analyzer for Sequence Typing & Fragment Analysis
Applied Biosystems™

3500xL Genetic Analyzer for Sequence Typing & Fragment Analysis

This 24-capillary Genetic Analyzer is specifically configured with software for both fragment analysis and sequence typing applications.
Have Questions?
Catalog NumberQuantity
44404711 system
Catalog number 4440471
Price (INR)
-
Quantity:
1 system

This 24-capillary Genetic Analyzer is specifically configured with software for both fragment analysis and sequence typing applications.

The 3500xL 24-capillary platform can run a wide variety of applications, including de novo sequencing and resequencing (mutational profiling), as well as microsatellite analysis, MLPA™, LOH, MLST, AFLP™ and SNP validation or screening. The majority of applications can be run on a single polymer and capillary array.

This package includes a 24 capillary instrument, the 3500 Data Collection Software, a Dell Workstation and monitor as well as reagent kits for system qualification. The product package also includes software for sequence typing and fragment analysis. The 3500 Series Data Collection Software integrates seamlessly with several downstream software packages to provide comprehensive analysis of genetic data.

Important Note:
Do you still have a 3100? 3100 Systems will be retired at the end of 2011. Take advantage of our limited time promotions to TRADE-IN your 3100 to get a brand new 3500. Please contact your local Sales Representative to learn more.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Amperage15 A
Capillary Length50 cm, 36 cm
Electrical Requirements100 V to 240 V, 50/60 HZ, 15 A
External ComputerHard Drive: 2x 500GB SATA 3.0 Gb/s and 8 MB Data Burst Cache, Memory: 16 GB (2x 8GB) 1600 MHz DDR3 Non-ECC. 4th Gen Intel Core I7 Processor- 3.1 GHz Turbo Processor, Operating System: Windows 7 SP1
For Use With (Equipment)POP-6™ Polymer for 3500⁄3500xL Genetic Analyzers, 3500xl Genetic Analyzer Capillary Array (50 cm), POP-4™ Polymer for 3500⁄3500xL Genetic Analyzers, POP-7™ Polymer for 3500⁄3500xL Genetic Analyzers, 3500xl Genetic Analyzer Capillary Array (36 cm)
Format96-well Plate, 8-tube Strip
Height (English) Exterior28.34 in.
Height (Metric) Exterior72 cm
Length (English) Exterior24.01 in.
Length (Metric) Exterior61 cm
Max. VoltageElectrophoresis Voltage - Up to 20 kV
Number of Capillaries24 Capillaries
Operating EnvironmentHumidity: 20%-80% (non-condensing), Room temp: <2° fluctuation during runs, Temperature: 15°C-30°C
Operating SystemWindows 7 (Service Pack 1)
PolymerPOP-4™, POP-6™, POP-7™
Polymer ConsumptionReplace at earliest of 7 days⁄960 samples⁄50 Injections (960-sample pouches), Replace at earliest of 7 days⁄384 samples⁄20 Injections (384-sample pouches)
Quantity1 system
Read Length≥850 bp using a 50 cm capillary and POP-7™
Software CompatibilitySequencing Analysis Software, GeneMapper™ Software
ThroughputUp to 1272 samples per day (using run module HID36 POP7)
Warranty1 Year Limited Warranty on Parts and Labor, Service Installation and Application Training Included
Weight82 kg (approximately)
Width (English) Exterior24.01 in.
Width (Metric) Exterior61 cm
Software TypeData Collection Software, Fragment Analysis
Starting MaterialDNA
Unit SizeEach
Package Components
4361142
Catalog Number
4361142
Unit Size
Qty
1
3500xL Genetic Analyzer
Catalog Number
4405633
Unit Size
1 system
Qty
1
4406933
Catalog Number
4406933
Unit Size
Qty
1
Contents & Storage
This package consists of the 24 capillary instrument, the 3500 Data Collection Software, a Dell Workstation and monitor as well as reagent kits for system qualification. This package also includes GeneMapper™ software for fragment analysis, and software for sequence typing applications.

Frequently asked questions (FAQs)

Is the Plate Septa, 96 well (Cat. No. 4315933) compatible with 3500/3500xL Genetic Analyzers?

Yes, it is compatible with all versions of 3500/3500xL Genetic Analyzers as well as the 3500/3500xL Dx Genetic Analyzers, specifically in RUO mode.

Is there a way to automatically upload my data into the Thermo Fisher Cloud app directly from the Applied Biosystems 3500 Genetic Analyzer, instead of using Data Collection Software?

If you are using the most recent version of Data Collection Software (v3.3), there is a way to automatically upload your data into the Thermo Fisher Cloud app from the instrument. When a run is complete, the data is stored in the results group folder that is specified in the plate record. The data will be automatically uploaded to your account if your User ID is the active ID. Results are saved in your account folders named by the plate and injections, not in the results group folder. Please go to page 43 of the 3500/3500xL Genetic Analyzer User Guide for Data Collection Software v3.3 (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/100079380_3500v3_3RUO_UG.pdf) for details regarding the Thermo Fisher Cloud app.

If you are using Data Collection Software v3.1, you would have to manually upload your data into the Thermo Fisher Cloud app from your data folder on the computer.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.