Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh
Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh
Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh
Acros Organics

Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh

CAS: 13450-90-3 | Cl3Ga | 176.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
44410025025 gGlass bottle
Catalog number 444100250
HSN 28273990
Price (INR)
21,943.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
21,943.00
Each
Chemical Identifiers
CAS13450-90-3
IUPAC Namegallium(3+) trichloride
Molecular FormulaCl3Ga
InChI KeyUPWPDUACHOATKO-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ga+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Beads
Total trace metal impurities=<100 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only