Custom Plus TaqMan™ RNA Assay, FAM
Applied Biosystems™

Custom Plus TaqMan™ RNA Assay, FAM

Applied Biosystems Custom Plus TaqMan RNA assays, are the custom version of our predesigned TaqMan assays for quantitative real-time PCR.Read more
Have Questions?
Catalog NumberQuantity
4441118L (2900 reactions), made to order
Catalog number 4441118
Price (INR)
150,283.00
Each
Quantity:
L (2900 reactions), made to order

Applied Biosystems Custom Plus TaqMan RNA assays, are the custom version of our predesigned TaqMan assays for quantitative real-time PCR. Custom Plus TaqMan RNA assays consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5’ end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3’ end.

Features include:
Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
Accurate—identify small fold-changes with high accuracy of quantitation
Custom content—easily design an assay for over 20 species
Designed using the full bioinformatic power of the TaqMan Assay design pipeline, which uses proprietary algorithms that have generated millions of assay designs and is regarded as the benchmark in the industry for qPCR design

Approximate ship time
Made to order: 4–6 days in North America, 6–10 days in Europe

Pre-formulated assay
• 2 unlabeled PCR primers (900 nM each final 1X concentration)
• 1 FAM dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Recommended master mix: TaqMan Fast Advanced Master Mix

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
No. of Reactions2900 reactions, made to order
Product LineTaqMan
QuantityL (2900 reactions), made to order
Shipping ConditionRoom Temperature
Concentration60X
Unit SizeEach
Contents & Storage
1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Frequently asked questions (FAQs)

Can I set up and store the TaqMan Fast Universal PCR Master Mix at room temperature if the instrument is in use?

For optimal results, we recommend to run the reaction plate right after setting up the reaction. If a reaction plate cannot be run within 2 hours of completing the reaction setup, then freeze/refrigerate the reaction plate until it can be loaded and run on the fast PCR instrument platform.

Can I run a reaction made with the standard TaqMan Universal PCR Master Mix in fast thermal cycling mode?

No, we do not recommend this approach. Since the standard TaqMan Universal PCR Master Mix is not optimized for running with fast thermal cycling, assay results will be significantly compromised.

Is the TaqMan Fast Universal PCR Master Mix offered with AmpErase UNG?

The TaqMan Fast Universal PCR Master Mix is provided without any UNG and you should add in AmpEraseUNG to reactions separately, with the addition of an initial 2 minute cycling step at 50º C. If you prefer to have a master mix with the UNG pre-added, please use TaqMan Fast Advanced Master Mix.

Can I compare performance of assays between the TaqMan Fast Universal PCR Master Mix compare and TaqMan Universal PCR Master Mix?

In general, assays have been found to perform comparably with these reagents, but they may not provide exactly identical results.

What are the storage conditions and the shelf life of the TaqMan Fast Universal PCR Master Mix?

We recommend a storage temperature of 4C and the shelf life is 9 months from the manufacturing date.