Product Image
Applied Biosystems™
TaqMan™ Non-Coding RNA Assay
TaqMan® Non-coding RNA Assays provide a highly reliable way to measure expression of long non-coding transcripts. These assays are basedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
4441122Extra small scale
4441120Small scale
Catalog number 4441122
Price (INR)
-
Description:
Extra small scale
TaqMan® Non-coding RNA Assays provide a highly reliable way to measure expression of long non-coding transcripts. These assays are based on TaqMan® Assays technology to help provide the high specificity and sensitivity you need for quantification and validation studies.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionExtra small scale
Product LineTaqMan
Quantityextra small scale
FormLiquid
Unit SizeEach
Contents & Storage
Store in freezer (-5° to -30°C).