Diethyl Ether, technical, stabilized
Diethyl Ether, technical, stabilized
Diethyl Ether, technical, stabilized
Diethyl Ether, technical, stabilized
Acros Organics

Diethyl Ether, technical, stabilized

CAS: 60-29-7 | C4H10O | 74.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
444260050
also known as 44426
5 LMetal Drum
Catalog number 444260050
also known as 44426
HSN 29091100
Price (INR)
16,452.00
Each
Quantity:
5 L
Packaging:
Metal Drum
Request bulk or custom format
Price (INR)
16,452.00
Each
Chemical Identifiers
CAS60-29-7
IUPAC Nameethoxyethane
Molecular FormulaC4H10O
InChI KeyRTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILESCCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Density0.713 to 0.716 kg/l
Stabilizer5 to 10 ppm BHT
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only