Product Image
Applied Biosystems™
GeneMapper™ ID-X Software v1.2, Client Installation
This kit includes the GeneMapper™ ID-X v1.2 Client Installation (no database) with 5 licenses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
44432601 License
444325810 Licenses
Catalog number 4443260
Price (INR)
2,438,387.00
Each
Quantity:
1 License
Price (INR)
2,438,387.00
Each
This kit includes the GeneMapper™ ID-X v1.2 Client Installation (no database) with 5 licenses. Documentation included.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
For Use With (Equipment)310 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
Operating SystemWindows Vista, Windows 2000, Windows XP
Product TypeSoftware V1.2 Client Installation
Quantity1 License
Product LineGeneMapper
Software TypeForensic STR Fragment Analysis
Unit SizeEach