4-Amino-3-fluorophenylboronic acid pinacol ester, 97%
4-Amino-3-fluorophenylboronic acid pinacol ester, 97%
4-Amino-3-fluorophenylboronic acid pinacol ester, 97%
4-Amino-3-fluorophenylboronic acid pinacol ester, 97%
Acros Organics

4-Amino-3-fluorophenylboronic acid pinacol ester, 97%

CAS: 819058-34-9 | C12H17BFNO2 | 237.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 444500050
also known as 44450
HSN 29319090
Price (INR)
137,176.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
137,176.00
Each
Chemical Identifiers
CAS819058-34-9
IUPAC Name2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC12H17BFNO2
InChI KeyAIXGNRNTXUKZLC-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Appearance (Color)Off-white to tan
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only