4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 97%
4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 97%
4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 97%
4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 97%
Acros Organics

4,4-Dimethyl-2-cyclohexen-1-one-2-boronic acid pinacol ester, 97%

CAS: 219489-09-5 | C14H23BO3 | 250.15 g/mol
Have Questions?
Catalog NumberQuantity
444980010
also known as 44498
1 g
Catalog number 444980010
also known as 44498
HSN 29319090
Price (INR)
65,168.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (INR)
65,168.00
Each
Add to cart
Chemical Identifiers
CAS219489-09-5
IUPAC Name4,4-dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohex-2-en-1-one
Molecular FormulaC14H23BO3
InChI KeyMMYLPQABFZSMCP-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(C)(C)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to yellow
Infrared spectrumConforms
Melting point98°C to 102°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only