4-Formylphenylboronic acid pinacol ester, 97%
4-Formylphenylboronic acid pinacol ester, 97%
4-Formylphenylboronic acid pinacol ester, 97%
Acros Organics

4-Formylphenylboronic acid pinacol ester, 97%

CAS: 128376-64-7 | C13H17BO3 | 232.09 g/mol
Have Questions?
Catalog NumberQuantity
445040250
also known as 44504
25 g
Catalog number 445040250
also known as 44504
HSN 29319090
Price (INR)
59,226.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
59,226.00
Each
Chemical Identifiers
CAS128376-64-7
IUPAC Name4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular FormulaC13H17BO3
InChI KeyDMBMXJJGPXADPO-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light yellow
Melting point58°C to 64°C
GC>=96.0 %
NMR (1H-NMR)Consistent with structure
Appearance (Form)Solid or crystalline solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only