n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.
Acros Organics

n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur.

CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Catalog NumberQuantity
4457200101 L
Catalog number 445720010
HSN 29011000
Price (INR)
7,267.00
Each
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Density(20/20°C) 0.659 to 0.663
GC>=98.5 % (Sum of 5 isomers, total hexanes,
Residue after evaporation=<0.0005 %
ThiophenePasses test
UVat 210 nm A: =<0.25
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only