Product Image
Invitrogen™

Ambion™ In Vivo GAPDH Positive Control siRNA

Ambion™ In Vivo siRNAs are designed using the proven Silencer™ Select algorithm and incorporate chemical modifications for superior serum stability with in vivo delivery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
44572885 nmol
445940750 nmol
4457291250 nmol
Catalog number 4457288
Price (INR)
25,713.00
Online offer
Ends: 31-Dec-2026
42,854.00
Save 17,141.00 (40%)
Each
Quantity:
5 nmol
Price (INR)
25,713.00
Online offer
Ends: 31-Dec-2026
42,854.00
Save 17,141.00 (40%)
Each

Ambion™ In Vivo siRNA Controls

  • Validated siRNA controls for optimizing siRNA experiments targeting coding and noncoding transcripts
  • Positive Control siRNAs functionally tested in several common cell lines
  • Negative Controls functionally proven to have minimal effects on cell proliferation and viability
  • Include Silencer™ Select modifications for enhanced specificity or Ambion™ In Vivo modifications for increased stability
  • For use in human, mouse, and rat cells

What are Ambion™ In Vivo siRNAs?
Ambion™ In Vivo siRNAs are designed using the proven Silencer™ Select algorithm and incorporate chemical modifications for superior serum stability with in vivo delivery. The added serum stability does not compromise the performance of the siRNAs. Ambion™ In Vivo siRNAs have been shown to be non-toxic in vivo (mouse) and non-immunogenic (cell-based assays). In cell-based assays, Ambion™ In Vivo siRNAs exhibit equivalent or better potency than Silencer™ Select siRNAs. The result is better confidence in your in vivo RNAi experiments, better knockdown when paired with an efficient delivery solution, and the assurance of limited non-specific effects.

GAPDH Positive Control siRNAs
Our extensively validated, positive control siRNA to human, mouse, and rat GAPDH serves multiple functions. First, it is an ideal 'test' siRNA for those just beginning siRNA experiments, because it is validated to work in multiple cell lines. In addition, because it targets GAPDH mRNA, which is commonly used as an internal control, its effects are easy to assay, and thus provides an excellent tool to monitor siRNA transfection efficiency by real-time RT-PCR. The Ambion™ In Vivo GAPDH Positive Control siRNA shows increased serum and nuclease resistance compared with Silencer™ Select siRNAs without loss of potency or specificity and is recommended for in vivo applications.

Quality Control
Ambion™ synthesizes and purifies each Ambion™ In Vivo siRNA in state-of-the-art facilities to meet the highest quality standards. As part of our rigorous quality control procedures, each RNA oligonucleotide is analyzed by MALDI-TOF mass spectrometry, and analytical HPLC is used to monitor purity. To provide the utmost in quality, we also assess each annealed siRNA by gel electrophoresis to confirm that the strands anneal properly. The result is premium-quality siRNA that is purified and ready to use.

In Vivo Ready Controls
Our 'In Vivo Ready' Control siRNAs are subjected to that extra level of purification and testing required for the introduction of siRNAs into animals. After HPLC purification and annealing, each siRNA is further purified utilizing a process that removes excess salt via a semi-permeable membrane. The result is highly pure siRNA with minimal salt content, suitable for in vivo applications. In vivo siRNAs are then sterile filtered, and tested for the presence of endotoxin. At concentrations of 50 μM in presence of deionized water, in vivo Ready siRNAs contain less than 5.0 mM Na+, less than 0.06 mM K+, and less than 0.02 mM Mg2+.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)RNAi, Transfection
Label or DyeUnconjugated
Product LineAmbion
Product TypesiRNA
PurityIn Vivo, HPLC
Quantity5 nmol
Shipping ConditionRoom Temperature
Control TypePositive Control
RNAi TypesiRNA
TargetGAPDH
Unit SizeEach
Contents & Storage
The siRNA (annealed) is shipped at ambient temperature as dried down oligonucleotides in a single tube, along with 1.75 ml Nuclease-free Water for resuspension. Store at or below –20°C.

Frequently asked questions (FAQs)

What are the benefits of using our Invitrogen In vivo siRNA?

- 100x more stable than unmodified siRNA - longer knockdown in vivo
- Compatible with any 21-mer sequence - no loss of potency
- Non-toxic - actually reduces siRNA sequence specific immune responses
- Enhanced specificity - up to 90% reduction in off target effects
- Both Pre-designed and Validated Invitrogen In Vivo siRNAs offered


Find additional tips, troubleshooting help, and resources within our RNAi Support Center.

What siRNA do you offer that I can use for my in vivo studies, and what are the differences between them?

Please see the different siRNA we offer for in vivo studies:

Ambion In Vivo Pre-designed siRNAs are designed using the Silencer Select algorithm and incorporate additional chemical modifications for superior serum stability (half life >5 hours at 37 degrees C in 90% mouse serum) with in vivo applications. Ambion In Vivo siRNAs are non-toxic and non-immunogenic in vitro (peripheral blood nononuclear cells; PBMC) and in vivo (mouse). In cell-based assays, Ambion In Vivo siRNAs exhibit potency and specificity equivalent to Silencer Select siRNAs.
Ambion In Vivo Custom Designed siRNAs are designed to your specified target, using the Silencer Select algorithm and Ambion In Vivo chemical modifications.
Ambion In Vivo Custom Synthesized siRNAs are synthesized with your sequence, with Ambion In Vivo chemical modifications for serum stability.

Our Ambion In Vivo siRNA can be ordered with the following purities:

Desalted: Desalted and analyzed by MALDI-TOF mass spectrometry.
HPLC: HPLC purified and analyzed by MALDI-TOF and analytical HPLC
In Vivo Ready : HPLC purified, analyzed by MALDI-TOF and analytical HPLC, dialyzed to remove salts, sterile filtered, and endotoxin tested

Use our GeneAssist tool to order your custom siRNA (http://www.thermofisher.com/order/custom-genomic-products/tools/sirna/).

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.