Ammonium sulfate, 99%, pure
Ammonium sulfate, 99%, pure
Ammonium sulfate, 99%, pure
Acros Organics

Ammonium sulfate, 99%, pure

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
5 kg
Packaging:
View all
Fibre drum
Catalog number 446450250
also known as 44645
HSN 28332990
Price (INR)
39,839.00
Each
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (INR)
39,839.00
Each
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to almost white
Appearance (Form)Crystalline powder or crystals
Titration excess NaOH>=98.5 %
Loss on drying=<0.2 %
pH5 to 6 (5 % at 20°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only