Ultra Turrax™ Tube Drive from IKA™ for PGM™ System
Ion Torrent™

Ultra Turrax™ Tube Drive from IKA™ for PGM™ System

Have Questions?
Catalog NumberQuantity
44647471 Instrument
Catalog number 4464747
Price (INR)
-
Quantity:
1 Instrument
The ULTRA-TURRAX™ Tube Drive from IKA™ for the Personal Genome Machine™System is a sample preparation instrument for use in the Personal Genome Machine™ sequencing workflow. This customized benchtop device is used for the preparation of appropriately sized reactors for emulsion PCR. This device can be used in conjunction with Template Preparation Tubes.

The ULTRA-TURRAX™ Tube Drive from IKA™ for the PGM™ System provides you with:
• Lower cost per sample
• Scalability

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Amperage800 mA
For Use With (Application)Sequencing
For Use With (Equipment)Ion PGM™ System
OutputSemiconductor sequencing emulsion
Product LineIKA, Ion Torrent, Ultra-Turrax
Product TypeTube Drive
Quantity1 Instrument
Shipping ConditionRoom Temperature
Voltage24 V
Unit SizeEach
Contents & Storage
Contains one instrument.