PrepSEQ™ Express Nucleic Acid Extraction Kit
Applied Biosystems™

PrepSEQ™ Express Nucleic Acid Extraction Kit

The PrepSEQ Express Nucleic Acid Extraction Kit contains pre-filled reagents and plastic tubes and tips to perform hands-free extraction of purified nucleic acid from 52 samples.
Have Questions?
Catalog NumberQuantity
44663511 Kit
Catalog number 4466351
Price (INR)
-
Quantity:
1 Kit

The PrepSEQ Express Nucleic Acid Extraction Kit contains pre-filled reagents and plastic tubes and tips to perform hands-free extraction of purified nucleic acid from 52 samples.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Final Product TypeGenomic DNA, Total RNA
For Use With (Equipment)7500 Fast System, AutoMate Express™ Nucleic Acid Extraction System
High-throughput CompatibilityAutomated Protocols
No. of SamplesSufficient for isolation of 52 samples
PCR MethodReal-Time Quantitative PCR (qPCR)
Product LinePrepSEQ
Quantity1 Kit
Sample TypeBiopharmaceutical Purification Sample, Bulk Drug Substance Sample, Cell Cultures
Shipping ConditionRoom Temperature
TargetGenomic DNA, Total RNA
Test Type≤ 1 hr.
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
Contains pre-filled reagent cartridges and plastics to perform extraction of DNA and RNA.

Store at room temperature.