(R)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific Chemicals
(R)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific Chemicals
(R)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific Chemicals
Acros Organics

(R)-2-(Trifluoromethyl)piperidine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
446690010
also known as 44669
1 g
Catalog number 446690010
also known as 44669
HSN 29333919
Price (INR)
26,228.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
26,228.00
Each
Chemical Identifiers
CAS1212734-83-2
IUPAC Name2-(trifluoromethyl)piperidine
Molecular FormulaC6H10F3N
InChI KeyNAXDEFXCCITWEU-UHFFFAOYNA-N
SMILESFC(F)(F)C1CCCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+10° to +20° (20°C, 589 nm) (c=4 in methanol)
Appearance (Color)Clear colorless to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only