PrepFiler™ LySep™ Columns
Applied Biosystems™
PrepFiler™ LySep™ Columns
PrepFiler LySep Columns are intended for use with the PrepFiler Forensic DNA Extraction kits and allow users to easily perform lysis and substrate removal in a single tube assembly.
Have Questions?
Catalog NumberQuantity
4468323500 Columns
Catalog number 4468323
Price (INR)
415,516.00
Each
Quantity:
500 Columns
Price (INR)
415,516.00
Each
PrepFiler LySep Columns are intended for use with the PrepFiler Forensic DNA Extraction kits and allow users to easily perform lysis and substrate removal in a single tube assembly. There is no transfer of the substrate to another tube for removal, thus minimizing hands-on time and helping to reduce the risk of cross contamination and sample transposition events. The only analyst interaction and handling of the substrate occurs during the addition of the substrate to the PrepFiler LySep Column for lysis.
The innovative PrepFiler LySep Column dramatically streamlines the off-line lysis portion of the extraction method. PrepFiler Lysis Buffer or PrepFiler BTA Lysis buffer is added directly to the PrepFiler LySep Column along with the sample for incubation. Next, the PrepFiler LySep Column is briefly centrifuged, causing the lysate to flow through the burstable membrane while the substrate remains behind in the column. The sample tube is then ready to be placed on the AutoMate Express instrument. This eliminates the manual lysate and substrate transfer steps, helping save time and minimizing cross contamination and sample transposition events.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)AutoMate Express Forensic DNA Extraction System
Product LinePrepFiler
Product TypeDNA Extraction Column
Quantity500 Columns
Shipping ConditionRoom Temperature
FormatIndividual
Unit SizeEach