QuantStudio™ 12K Flex Real-Time PCR System, 384-well block, desktop
QuantStudio™ 12K Flex Real-Time PCR System, 384-well block, desktop
Applied Biosystems™

QuantStudio™ 12K Flex Real-Time PCR System, 384-well block, desktop

The QuantStudio™ 12K Flex system is a highly flexible, comprehensive real-time PCR platform that leverages the best of Applied BiosystemsRead more
Have Questions?
Catalog NumberQuantity
44711341 system
Catalog number 4471134
Price (INR)
-
Request A Quote
Quantity:
1 system
The QuantStudio™ 12K Flex system is a highly flexible, comprehensive real-time PCR platform that leverages the best of Applied Biosystems real-time technology in a single instrument. The QuantStudio™ 12K Flex Real-Time PCR System includes easy-to-change blocks that can accommodate OpenArray® plates, TaqMan® array cards, 384-well, 96-well, and FAST 96-well plates allowing you to select the format that is right for your project whether it’s now or in the future, for maximum flexibility and consistency in your research.

Thermo Fisher Cloud
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.

The QuantStudio™ 12K Flex System’s intuitive software, easy touch screen setup, automation integration, effortless block exchange, and easy-to- use OpenArray® components are designed to let you get started right away and reduce hands on time. Plan for the future and save time, money, and space in your lab with the only all-in-one qPCR instrument enabling a broad range of applications with maximum throughput.

Flexibility & Scalability
Includes five interchangeable blocks: The QuantStudio™ 12K Flex blocks accomodate 96-well, FAST 96-well, 384-well, TaqMan® array cards, and OpenArray® plates, for effortless scaling from 1 to 12,000 data points in a single run. Increase throughput as you dive deeper into your research, using just one instrument. As you begin your project, start with 96-well format experiments and then scale up to a higher throughput to get the statistically significant answers you need. Additionally, you can easily validate and screen your targets of interest with a selected panel of genes using novel formats such as the TaqMan® array cards and OpenArray® plates.

• Compatible with a full range of TaqMan® assays:
The QuantStudio™ 12K Flex System is optimized to enable clear, clean data for all TaqMan® assays that might be used across a broad range of applications, including gene expression, microRNA and small noncoding RNAs, SNP genotyping, protein analysis, protein thermal shift, high resolution melt, copy number variation, and pathogen detection.

• Provides a seamless switch from qPCR to digital PCR:
Increase the precision and sensitivity of your nucleic acid experiment by switching from qPCR to digital PCR mode using QuantStudio™ Digital PCR kits and DigitalSuite™ Software with the QuantStudio™ OpenArray® block.

• Delivers peace of mind:
Leave early on a Friday and monitor runs from home, knowing that the system can safely store over 100 runs.

Maximum Throughput
Enhanced OptiFlex® System: Building upon Applied Biosystems’ proprietary technology, the Enhanced OptiFlex® System provides enhanced fluorescence detection for the accurate and sensitive data analysis necessary to collect 12,000 data points in a single run.

• Unprecedented coverage and sample throughput:
The OpenArray® format accelerates genomic validation and screening, driving over 150,000 data points per work day, enabling most projects to be completed in days instead of weeks.

• Simple OpenArray® workflow:
QuantStudio™ Open Array® plates arrive in a case to facilitate ease of handling and ensure optimal data quality upon loading of samples using the Accufill™ System. Easily load four arrays into the QuantStudio™ 12K Flex instrument to achieve over 12,000 data points. Integrated sample tracking software lets you easily track your samples throughout the course of your project.

• Integrated analysis and quality systems:
New intuitive touch-screen interface and integrated calendar feature allows for scheduling time of use on the instrument. Additional analysis software such as DigitalSuite™ v1.0, ExpressionSuite v1.0, and Genotyper v1.1 is available to help you analyze hundreds of targets and samples and achieve the statistical power you need.

QuantStudio™ 12K Flex Software
• Intuitive interface and innovative design for novice and experienced users
• Convenient walk-away operation
• Improved data analysis across block types
• Fully compatible with high-throughput environments

Expandable Software Architecture
Optional add-ons are available to upgrade the existing software:
21 CFR Part 11 compliance: This optional SAE module assists with Security, Auditing, and Electronic signature of records for full data traceability.

• High Resolution Melt (HRM):
Using the built-in MeltDoctor™ protocols and calibrations, you can spend less time optimizing the run and more time customizing your results. Define the number of variants you want to see by quickly changing analysis settings.

• DigitalSuite™ Software v1.0:
For OpenArray® block users needing sensitivity for rare targets and precision to measure small differences in target abundance.

• ExpressionSuite Software v1.0:
our latest stand-alone gene expression software with enhanced analysis tools that can analyze over 200 384-well plates or 10 OpenArray® plates.

Learn more about available financing options and programs ›

Specifications
Detection MethodSYBR, Primer-Probe Detection
Dimensions51 cm (W) x 74 cm (H) x 67 cm (D)
Display TypeTouchscreen
Dynamic RangeUp to 9 Logs of Linear Dynamic Range
Electrical Requirements60 Hz
External ComputerComputer and Monitor
For Use With (Equipment)QuantStudio™
Format384-well Plate
High-throughput CompatibilityHigh-throughput Compatible, Multiplexing
Optics6 Emission Filters (500-720 nm), 6 Excitation Filters (450-670 nm), 21 Color Options
Other Format Options96-well Fast Block Upgrade Option, OpenArray™ Block Upgrade Option, TaqMan™ Array Card Block Upgrade Option, 96-well Standard Block Upgrade Option
Product LineQuantStudio
ProgramsRemote Monitoring of up to 10 Instruments
Quantity1 system
Reaction SpeedStandard
Reaction Volume Range5–20 μL (384-well Plate)
SensitivityDown to 1 Copy
Voltage100 to 240 V
Weight69 kg (152 lb.)
Block FormatInterchangeable
Unit SizeEach
Contents & Storage
Desktop Computer

Frequently asked questions (FAQs)

What is the difference between continuous melt curves and step and hold melt curves on the Applied Biosystems Real-Time PCR Instruments?

Continuous melt curve: increases the temperature by the ramp increment (degrees C/sec) so that the temperature will change at a constant rate set in the protocol.
Step and Hold melt curve: increases the temperature by the ramp increment (degrees C) and then holds at that temperature for the time specified by the user.

The amount of data collected will depend on the filter(s) selected. The more filters you use, the longer the acquisition will take and the fewer data points will be collected.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

I recently switched the block in my real-time PCR instrument. Now it shows the status as 'not calibrated' for all the calibration plates, even though my calibrations are current. What is causing this issue?

The issue might be due to the block not being properly seated in the instrument. Please follow the steps below:

  1. Remove the block from the instrument.
  2. Reinsert the block into the instrument.


If the problem persists after reseating the block, please contact our Technical Support team for further assistance.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio 12K Flex Software?

If you are analyzing results from the QuantStudio 12K Flex Real-Time PCR System, it is possible to import a standard curve into the QuantStudio 12K Flex Software following the instructions below:
1. Open the data file in the QuantStudio 12K Flex Software.
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply Analysis Settings.

What is the difference between a Fast 96-well and standard 96-well block?

There are two main differences between a Fast 96-well and standard 96-well block for the Applied Biosystems real-time PCR systems:
• Reaction well volume: The Fast 96-well blocks have 0.1 mL reaction wells and are compatible with 0.1 mL plates and tubes. The standard 96-well blocks have 0.2 mL reaction wells and are compatible with 0.2 mL plates and tubes.
• Ramp rate: The fast 96-well blocks have a higher maximum ramp rate than standard blocks for the same real-time PCR system. However, both the fast 96-well and standard 96-well blocks are capable of running standard and fast chemistry.

Note: For newer instruments (QuantStudio 3 and 5 Real-Time PCR Systems and later) "0.1 mL" and "0.2 mL" are being used in place of "fast" and "standard" as designations for the two 96-well block formats.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.