Acetic acid, ammonium salt, 98%, for biochemistry
Acetic acid, ammonium salt, 98%, for biochemistry
Acetic acid, ammonium salt, 98%, for biochemistry
Acros Organics

Acetic acid, ammonium salt, 98%, for biochemistry

CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol
Have Questions?
Catalog NumberQuantityPackaging
447125000
also known as 44712
500 gPlastic bottle
Catalog number 447125000
also known as 44712
HSN 29152990
Price (INR)
9,469.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
9,469.00
Each
Chemical Identifiers
CAS631-61-8
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<2 ppm
UV(1 M in water)
UVat 254 nm A: =<0.02
UVat 280 nm A: =<0.01
UVat 350 nm A: =<0.01
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only