Product Image
Applied Biosystems™
MagMax™ Express-96 PCR Well Magnetic Head
This is a magnetic head for the MagMAX™ Express-96 Deep Well Magnetic Particle Processor that is designed for use withRead more
Have Questions?
Catalog NumberQuantity
44729911 head
Catalog number 4472991
Price (INR)
973,571.00
Each
Quantity:
1 head
Price (INR)
973,571.00
Each
This is a magnetic head for the MagMAX™ Express-96 Deep Well Magnetic Particle Processor that is designed for use with PCR plates. Includes one magnetic head. Not compatible with the KingFisher™ Flex 96 Magnetic Particle Processor.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 head
Shipping ConditionRoom Temperature
Product LineMagMAX
TypeMagnetic Head
Unit SizeEach