TaqMan™ miRNA ABC Purification Kit - Human Panel A
Applied Biosystems™

TaqMan™ miRNA ABC Purification Kit - Human Panel A

Use the TaqMan™ miRNA ABC Purification Kit to purify high-quality miRNA from blood-related or other difficult sample types without hazardousRead more
Have Questions?
Catalog NumberQuantity
447308740 Preps
Catalog number 4473087
Price (INR)
108,994.00
Each
Quantity:
40 Preps
Price (INR)
108,994.00
Each
Use the TaqMan™ miRNA ABC Purification Kit to purify high-quality miRNA from blood-related or other difficult sample types without hazardous chemicals.
• Simple and fast workflow using oligonucleotide-conjugated Dynal™ magnetic beads
• Small sample input: typical volumes range from 10 μL for blood samples to 50 μL for serum and plasma samples
• RT-PCR inhibitors commonly found in blood-related samples are easily washed away
• No need for hazardous chemicals such as phenol, chloroform, Trizol™ reagent, or guanidine isothiocyanate

Simple and Fast miRNA Purification
The TaqMan™ miRNA ABC (anti-miRNA bead capture) purification kit contains buffers and magnetic beads for single-tube isolation of specific microRNA (miRNA) from small inputs of all human sample types, including blood, serum, plasma, FFPE samples, solid tissues, cultured cells, and saliva in 75 minutes (see workflow, Figure 1).

Human Panel A and Human Panel B
The TaqMan™ miRNA ABC Purification Kit is offered in two configurations: Human Panel A and Human Panel B. The beads in each kit are superparamagnetic Dynabeads™ covalently bound to a unique set of 377 anti-miRNA oligonucleotides for each panel that correspond to the assays contained in the TaqMan™ Array Human MicroRNA A and B cards. The miRNA isolation process relies on hybridization of miRNAs to the corresponding anti-miRNA oligonucleotides attached to the beads. Complete lists of miRNAs isolated by the panels can be found in the TaqMan™ miRNA ABC Purification Kit protocol (below).

miRNA Analysis
The miRNAs isolated using the TaqMan™ miRNA ABC Purification Kit are ready for conversion into cDNA and downstream analysis by quantitative PCR using either single tube (individual) TaqMan™ microRNA assays or the TaqMan™ Array Human microRNA A and B cards. Figure 2 gives examples of the data that can be obtained from miRNA isolated from various sample types using the TaqMan™ miRNA ABC Purification Kit.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product Typemicro RNA
For Use With (Application)Real-Time Quantitative PCR (qPCR), microRNA Analysis
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product LineTaqMan
Product TypemiRNA ABC Purification Kit
Purification Time75 min.
Quantity40 Preps
System TypeDynabeads™
FormatKit
Isolation TechnologyMagnetic Bead
RNAi TypemiRNA
Sample TypeCell Cultures, Plasma, Saliva, Serum, Tissue, Tissue (FFPE), Tissue Sections, Urine, Whole Blood
SpeciesHuman
TargetmiRNA
Unit SizeEach
Contents & Storage
Buffer Kit - Store at room temperature
Lysis Buffer
ABC Buffer
Wash Buffer 1
Wash Buffer 2
Elution Buffer
1.5 mL LoBind tubes

Human Panel A Beads
Store at 2 °C to 8 °C

Frequently asked questions (FAQs)

I am using the TaqMan miRNA ABC Purification Kit and am having difficulty detecting miRNA in qPCR assays. What is the issue?

Here are possible causes and solutions:

- RNA degradation due to RNAse contamination during procedure: See “Guidelines to prevent RNAse contamination” on page 12 of the manual (https://tools.thermofisher.com/content/sfs/manuals/4473439C.pdf) for proper handling of sample.
- Inefficient elution of miRNA from the beads: follow hybridization, wash, and elution steps on pages 17-18.
- Inefficient washing: fully resuspend beads in wash buffers, and completely remove buffers at the end of the washes.
- Beads dried out during procedure: prepare the beads just before use and keep the beads suspended in liquid and the tube cap closed in between steps.
- Buffers not diluted as directed: prepare buffers as directed on page 13.
- Kit components compromised due to inappropriate storage conditions: store reagents as directed in “Kit contents and storage conditions” on page 9 of the manual.