TaqMan® OpenArray® Genotyping Barcode Panel 32A
Actual product may vary
Applied Biosystems™
TaqMan® OpenArray® Genotyping Barcode Panel 32A
Have Questions?
Catalog number 4475366
Price (INR)
-
The TaqMan® OpenArray® Genotyping Barcode Panel 32A consists of 3 Y-markers for gender confirmation and 29 autosomal SNPs that allow for unique identification of individual samples for biorepository applications. Panel 32B (available separately) is an additional set of autosomal SNPs that can be used if more SNPs are needed for increased stringency. The combined panels provide 3 Y-markers and 61 autosomal SNPs, allowing a total of 95 samples and one no template control to be run.
Markers were selected based on high minor allele frequency in multiple ethnic populations, representation on high throughput genotyping, and robust genotyping performance. These markers allow unique identification of individuals within sample collections. The panel allows unique identification of individuals throughout experimental processes by providing both sample identity and gender checks in the event of sample mix-ups or labeling mistakes. It also serves as an internal control for genome-wide association studies by containing markers from large microarrays to allow matching of sample genotypes directly from the panel to a GWAS array.
This panel is intended for use with the TaqMan® OpenArray® Real-time PCR System, which can cycle and image 3 plates simultaneously, providing high-throughput results with ease.
Note: equivalent panels for use with the QuantStudio™ 12K Flex instrument are also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size1 plate
Contents & Storage
Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C