TaqMan™ OpenArray™ Genotyping Barcode Panel 60, QuantStudio™ 12K Flex plate
Applied Biosystems™

TaqMan™ OpenArray™ Genotyping Barcode Panel 60, QuantStudio™ 12K Flex plate

The TaqMan OpenArray Genotyping Barcode Panel 60 consists of 3 Y-markers and 57 autosomal SNPs that allow for unique identificationRead more
Have Questions?
Catalog NumberQuantity
44753941 plate
Catalog number 4475394
Price (INR)
107,418.00
Each
Quantity:
1 plate
Price (INR)
107,418.00
Each
The TaqMan OpenArray Genotyping Barcode Panel 60 consists of 3 Y-markers and 57 autosomal SNPs that allow for unique identification of individual samples for biorepository applications. A total of 47 samples and 1 no template control can be run with this panel.

Markers were selected based on high minor allele frequency in multiple ethnic populations, representation on high throughput genotyping, and robust genotyping performance. These markers allow unique identification of individuals within sample collections. The panel allows unique identification of individuals throughout experimental processes by providing both sample identity and gender checks in the event of sample mix-ups or labeling mistakes. It also serves as an internal control for genome-wide association studies by containing markers from large microarrays to allow matching of sample genotypes directly from the panel to a GWAS array.

This panel is intended for use with the QuantStudio 12K Flex Real-time PCR system which can cycle and image 4 pates simultaneously, providing high-throughput results with ease.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
For Use With (Application)SNP Genotyping (Real-Time PCR-Based)
For Use With (Equipment)QuantStudio™ 12k Flex
Genotyping TargetSNPs (Known)
IncludesOpenarray Plate
Product LineOpenArray, TaqMan
Quantity1 plate
Sample TypeDNA
TypeGenotyping Barcode Panel
FormatOpenArray Plate
Unit SizeEach
Contents & Storage
Includes one OpenArray Plate containing a defined panel of pre-loaded, bench-validated TaqMan assays.
Store at -20 °C

Frequently asked questions (FAQs)

What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.