Product Image
Applied Biosystems™

PrepFiler™ 96-Well Spin Plates and Filter Plates, ethylene oxide-treated

Have Questions?
Catalog NumberQuantity
447603110 plate assemblies
Catalog number 4476031
Price (INR)
245,396.00
Each
Quantity:
10 plate assemblies
Price (INR)
245,396.00
Each
PrepFiler 96-Well Spin Plates and Filter Plates are intended for use with the updated PrepFiler Automated Forensic DNA Extraction Kit, which does not include plastic consumables, providing users the flexibility to order only the plastics consumables they need. PrepFiler Forensic DNA Extraction kits use magnetic particles with an optimized multi-component surface chemistry to deliver superior DNA yield from most forensic sample types, including body fluids, and stains and swabs of body fluids. The PrepFiler Spin Plates and Filter Plates are used for sample lysis and substrate removal for sample processing in 96-well plate format. The packaged plates are treated with ethylene oxide.
Specifications
For Use With (Application)Sequencing
No. of Reactions10 Assays
Product LinePrepFiler
Product Type96-Well Spin Plates and Filter Plate Set
Quantity10 plate assemblies
Shipping ConditionRoom Temperature
Format96-well Plate
Unit SizeEach
Contents & Storage
10 pre-filter/deep-well plate assemblies