Magnesium chloride hexahydrate, 98+%, extra pure
Magnesium chloride hexahydrate, 98+%, extra pure
Magnesium chloride hexahydrate, 98+%, extra pure
Acros Organics

Magnesium chloride hexahydrate, 98+%, extra pure

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number 447700050
also known as 44770
HSN 28273100
Price (INR)
25,731.00
Each
Quantity:
5 kg
Request bulk or custom format
Chemical Identifiers
CAS7791-18-6
IUPAC Namemagnesium(2+) hexahydrate dichloride
Molecular FormulaCl2H12MgO6
InChI KeyDHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Water51.0 to 55.0 %
Appearance (Form)Crystalline powder
Titration Complexometric98.0 to 101.0 %
Heavy metals (as Pb)=<10 ppm
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only